Learn More
enQuireBio™ Recombinant Human Proteasome subunit beta type-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6545-ec-100ug
Additional Details : Peso : 0.01000kg
Specifications
NP_002786.2 | |
Proteasome subunit beta type-3 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5691 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
PSMB3 | |
Recombinant Protein | |
E. coli | |
SIMSYNGGAV MAMKGKNCVA IAADRRFGIQ AQMVTTDFQK IFPMGDRLYI GLAGLATDVQ TVAQRLKFRL NLYELKEGRQ IKPYTLMSMV ANLLYEKRFG PYYTEPVIAG LDPKTFKPFI CSLDLIGCPM VTDDFVVSGT CAEQMYGMCE SLWEPNMDPD HLFETISQAM LNAVDRDAVS GMGVIVHIIE KDKITTRTLK ARMD |