missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prohibitin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Prohibitin-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8027-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001138303.1 | |
Prohibitin-2 Protein | |
Research Use Only | |
PHB2 | |
Recombinant Protein | |
E. coli | |
AQNLKDLAGR LPAGPRGMGT ALKLLLGAGA VAYGVRESVF TVEGGHRAIF FNRIGGVQQD TILAEGLHFR IPWFQYPIIY DIRARPRKIS SPTGSKDLQM VNISLRVLSR PNAQELPSMY QRLGLDYEER VLPSIVNEVL KSVVAKFNAS QLITQRAQVS LLIRRELTER AKDFSLILDD VAITELSFSR EYTAAVEAKQ VAQQEAQRAQ FLVEKAKQEQ RQKIVQAEGE AEAAKMLGEA LSKNPGYIKL RKIRAAQNIS KTIATSQNRI YLTADNLVLN LQDESFTRGS DSLIKGKK | |
Greater than 80% as determined by SDS-PAGE. |
11331 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |