missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Profilin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Profilin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8469-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005013.1 | |
Profilin-1 Protein | |
Research Use Only | |
PFN1 | |
Recombinant Protein | |
E. coli | |
AGWNAYIDNL MADGTCQDAA IVGYKDSPSV WAAVPGKTFV NITPAEVGVL VGKDRSSFYV NGLTLGGQKC SVIRDSLLQD GEFSMDLRTK STGGAPTFNV TVTKTDKTLV LLMGKEGVHG GLINKKCYEM ASHLRRSQY | |
Greater than 90% as determined by SDS-PAGE. |
5216 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |