missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prestin Protein
A DNA sequence encoding the Homo sapiens (Human) Prestin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9401-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001161434.1 | |
Prestin Protein | |
Research Use Only | |
SLC26A5 | |
Recombinant Protein | |
Yeast | |
YRTQSPSYKV LGKLPETDVY IDIDAYEEVK EIPGIKIFQI NAPIYYANSD LYSNALKRKT GVNPAVIMGA RRKAMRKYAK EVGNANMANA TVVKADAEVD GEDATKPEEE DGEVKYPPIV IKSTFPEEMQ RFMPPGDNVH TVILDFTQVN FIDSVGVKTL AGIVKEYGDV GIYVYLAGCS AQVVNDLTRN RFFENPALWE LLFHSIHDAV LGSQLREALA EQEASAPPSQ EDLEPNATPA TPEA | |
Greater than 90% as determined by SDS-PAGE. |
375611 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |