missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPME1 Protein
A cDNA sequence encoding the PPME1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13121-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
51400 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSALEKSMHL GRLPSRPPLP GSGGSQSGAK MRMGPGRKRD FSPVPWSQYF ESMEDVEVEN ETGKDTFRVY KSGSEGPVLL LLHGGGHSAL SWAVFTAAII SRVQCRIVAL DLRSHGETKV KNPEDLSAET MAKDVGNVVE AMYGDLPPPI MLIGHSMGGA IAVHTASSNL VPSLLGLCMI DVVEGTAMDA LNSMQNFLRG RPKTFKSLEN AIEWSVKSGQ IRNLESARVS MVGQVKQCEG ITSPEGSKSI VEGIIEEEEE DEEGSESISK RKKEDDMETK KDHPYTWRIE LAKTEKYWDG WFRGLSNLFL SCPIPKLLLL AGVDRLDKDL TIGQMQGKFQ MQVLPQCGHA VHEDAPDKVA EAVATFLIRH RFAEPIGGFQ CVFPGC | |
Greater than 95% as determined by SDS-PAGE. |
PPME1 Protein | |
Research Use Only | |
PPME1 | |
Human | |
His | |
The PPME1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 20% glycerol. |