missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POU domain class 2-associating factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) POU domain class 2-associating factor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7577-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006226.2 | |
POU domain class 2-associating factor 1 Protein | |
Research Use Only | |
POU2AF1 | |
Recombinant Protein | |
E. coli | |
MLWQKPTAPE QAPAPARPYQ GVRVKEPVKE LLRRKRGHAS SGAAPAPTAV VLPHQPLATY TTVGPSCLDM EGSVSAVTEE AALCAGWLSQ PTPATLQPLA PWTPYTEYVP HEAVSCPYSA DMYVQPVCPS YTVVGPSSVL TYASPPLITN VTTRSSATPA VGPPLEGPEH QAPLTYFPWP QPLSTLPTST LQYQPPAPAL PGPQFVQLPI SIPEPVLQDM EDPRRAASSL TIDKLLLEEE DSDAYALNHT LSVEGF | |
Greater than 90% as determined by SDS-PAGE. |
5450 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |