missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Potassium-transporting ATPase subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Potassium-transporting ATPase subunit beta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5692-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000696.1 | |
Potassium-transporting ATPase subunit beta Protein | |
Research Use Only | |
ATP4B | |
Recombinant Protein | |
Yeast | |
CLYVLMQTVD PYTPDYQDQL RSPGVTLRPD VYGEKGLEIV YNVSDNRTWA DLTQTLHAFL AGYSPAAQED SINCTSEQYF FQESFRAPNH TKFSCKFTAD MLQNCSGLAD PNFGFEEGKP CFIIKMNRIV KFLPSNGSAP RVDCAFLDQP RELGQPLQVK YYPPNGTFSL HYFPYYGKKA QPHYSNPLVA AKLLNIPRNA EVAIVCKVMA EHVTFNNPHD PYEGKVEFKL KIEK | |
Greater than 90% as determined by SDS-PAGE. |
496 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |