missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PNPT1 Protein
A cDNA sequence encoding the PNPT1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13086-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
87178 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
PNPT1 | |
Human | |
His | |
PNPT1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
PNPT1 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAVAVDLG NRKLEISSGK LARFADGSAV VQSGDTAVMV TAVSKTKPSP SQFMPLVVDY RQKAAAAGRI PTNYLRREIG TSDKEILTSR IIDRSIRPLF PAGYFYDTQV LCNLLAVDGV NEPDVLAING ASVALSLSDI PWNGPVGAVR IGIIDGEYVV NPTRKEMSSS TLNLVVAGAP KSQIVMLEAS AENILQQDFC HAIKVGVKYT QQIIQGIQQL VKETGVTKRT PQKLFTPSPE IVKYTHKLAM ERLYAVFTDY EHDKVSRDEA VNKIRLDTEE QLKEKFPEAD PYEIIESFNV VAKEVFRSIV LNEYKRCDGR DLTSLRNVSC EVDMFKTLHG SALFQRGQTQ VLCTVTFDSL ESGIKSDQVI TAINGIKDKN FMLHYEFPPY ATNEIGKVTG LNRRELGHGA LAEKALYPVI PRDFPFTIRV TSEVLESNGS SSMASACGGS LALMDSGVPI SSAVAGVAIG LVTKTDPEKG EIEDYRLLTD ILGIEDYNGD MDFKIAGTNK GITALQADIK LPGIPIKIVM EAIQQASVAK KEILQIMNKT ISKPRASRKE NGPVVETVQV PLSKRAKFVG PGGYNLKKLQ AETGVTISQV DEETFSVFAP TPSAMHEARD FITEICKDDQ EQQLEFGAVY TATITEIRDT GVMVKLYPNM TAVLLHNTQL DQRKIKHPTA LGLEVGQEIQ VKYFGRDPAD GRMRLSRKVL QSPATTVVRT LNDRSSIVMG EPISQSSSNS Q |