missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLEKHF2 Protein
A DNA sequence encoding the Homo sapiens (Human) PLEKHF2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8198-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_078889.1 | |
PLEKHF2 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
79666 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
PLEKHF2 | |
Recombinant Protein | |
E. coli | |
MVDRLANSEA NTRRISIVEN CFGAAGQPLT IPGRVLIGEG VLTKLCRKKP KARQFFLFND ILVYGNIVIQ KKKYNKQHII PLENVTIDSI KDEGDLRNGW LIKTPTKSFA VYAATATEKS EWMNHINKCV TDLLSKSGKT PSNEHAAVWV PDSEATVCMR CQKAKFTPVN RRHHCRKCGF VVCGPCSEKR FLLPSQSSKP VRICDFCYDL LSAGDMATCQ PARSDSYSQS LKSPLNDMSD DDDDDDSSD |