missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PHOSPHO2 Protein
A cDNA sequence encoding the PHOSPHO2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13044-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PHOSPHO2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMKILLV FDFDNTIIDD NSDTWIVQCA PNKKLPIELR DSYRKGFWTE FMGRVFKYLG DKGVREHEMK RAVTSLPFTP GMVELFNFIR KNKDKFDCII ISDSNSVFID WVLEAASFHD IFDKVFTNPA AFNSNGHLTV ENYHTHSCNR CPKNLCKKVV LIEFVDKQLQ QGVNYTQIVY IGDGGNDVCP VTFLKNDDVA MPRKGYTLQK TLSRMSQNLE PMEYSVVVWS SGVDIISHLQ FLIKD | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PHOSPHO2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |