missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Perilipin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Perilipin-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7848-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001073869.1 | |
Perilipin-4 Protein | |
Research Use Only | |
PLIN4 | |
Recombinant Protein | |
Yeast | |
GVDTSKTVLT GTKDTVCSGV TGAMNVAKGT IQTGVDTTKT VLTGTKNTVC SGVTGAVNLA KEAIQGGLDT TKSMVMGTKD TMSTGLTGAA NVAKGAMQTG LNTTQNIATG T | |
Greater than 90% as determined by SDS-PAGE. |
729359 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |