missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peptidyl-tRNA hydrolase ICT1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Peptidyl-tRNA hydrolase ICT1, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7530-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001290194.1 | |
Peptidyl-tRNA hydrolase ICT1, mitochondrial Protein | |
Research Use Only | |
ICT1 | |
Recombinant Protein | |
E. coli | |
LHKQKDGTEF KSIYSLDKLY PESQGSDTAW RVPNGAKQAD SDIPLDRLTI SYCRSSGPGG QNVNKVNSKA EVRFHLATAE WIAEPVRQKI AITHKNKINR LGELILTSES SRYQFRNLAD CLQKIRDMIT EASQTPKEPT KEDVKLHRIR IENMNRERLR QKRIHSAVKT SRRVDMD | |
Greater than 90% as determined by SDS-PAGE. |
3396 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |