Learn More
enQuireBio™ Recombinant Human Peptidyl-prolyl cis-trans isomerase-like 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Peptidyl-prolyl cis-trans isomerase-like 4, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP7976-ec-1mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 16051941
  • 2385.00€ / 1 miligramo
Fecha estimada de envío: 27-06-2024
para ver el stock
Especificaciones
NP_624311.1
Peptidyl-prolyl cis-trans isomerase-like 4 Protein
Research Use Only
PPIL4
Recombinant Protein
E. coli
MAVLLETTLG DVVIDLYTEE RPRACLNFLK LCKIKYYNYC LIHNVQRDFI IQTGDPTGTG RGGESIFGQL YGDQASFFEA EKVPRIKHKK KGTVSMVNNG SDQHGSQFLI TTGENLDYLD GVHTVFGEVT EGMDIIKKIN ETFVDKDFVP YQDIRINHTV ILDDPFDDPP DLLIPDRSPE PTREQLDSGR IGADEEIDDF KGRSAEEVEE IKAEKEAKTQ AILLEMVGDL PDADIKPPEN VLFVCKLNPV TTDEDLEIIF SRFGPIRSCE VIRDWKTGES LCYAFIEFEK EEDCEKAFFK MDNVLIDDRR IHVDFSQSVA KVKWKGKGGK YTKSDFKEYE KEQDKPPNLV LKDKVKPKQD TKYDLILDEQ AEDSKSSHSH TSKKHKKKTH HCSEEKEDED YMPIKNTNQD IYREMGFGHY EEEESCWEKQ KSEKRDRTQN RSRSRSRERD GHYSNSHKSK YQTDLYERER SKKRDRSRSP KKSKDKEKSK YR
Greater than 90% as determined by SDS-PAGE.
85313
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol