missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCNA Protein
A DNA sequence encoding the Homo sapiens (Human) PCNA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6470-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002583.1 | |
PCNA Protein | |
Research Use Only | |
PCNA | |
Recombinant Protein | |
E. coli | |
MFEARLVQGS ILKKVLEALK DLINEACWDI SSSGVNLQSM DSSHVSLVQL TLRSEGFDTY RCDRNLAMGV NLTSMSKILK CAGNEDIITL RAEDNADTLA LVFEAPNQEK VSDYEMKLMD LDVEQLGIPE QEYSCVVKMP SGEFARICRD LSHIGDAVVI SCAKDGVKFS ASGELGNGNI KLSQTSNVDK EEEAVTIEMN EPVQLTFALR YLNFFTKATP LSSTVTLSMS ADVPLVVEYK IADMGHLKYY LAPKIEDEEG S | |
Greater than 90% as determined by SDS-PAGE. |
5111 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |