missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human p53 / TP53 Protein
A DNA sequence encoding the Homo sapiens (Human) p53 / TP53, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6822-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000537.3 | |
p53 / TP53 Protein | |
Research Use Only | |
TP53 | |
Recombinant Protein | |
E. coli | |
MEEPQSDPSV EPPLSQETFS DLWKLLPENN VLSPLPSQAM DDLMLSPDDI EQWFTEDPGP DEAPRMPEAA PPVAPAPAAP TPAAPAPAPS WPLSSSVPSQ KTYQGSYGFR LGFLHSGTAK SVTCTYSPAL NKMFCQLAKT CPVQLWVDST PPPGTRVRAM AIYKQSQHMT EVVRRCPHHE RCSDSDGLAP PQHLIRVEGN LRVEYLDDRN TFRHSVVVPY EPPEVGSDCT TIHYNYMCNS SCMGGMNRRP ILTIITLEDS SGNLLGRNSF EVRVCACPGR DRRTEEENLR KKGEPHHELP PGSTKRALPN NTSSSPQPKK KPLDGEYFTL QIRGRERFEM FRELNEALEL KDAQAGKEPG GSRAHSSHLK SKKGQSTSRH KKLMFKTEGP DSD | |
Greater than 90% as determined by SDS-PAGE. |
7157 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |