missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human p53-regulated apoptosis-inducing protein 1
A DNA sequence encoding the Homo sapiens (Human) p53-regulated apoptosis-inducing protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8199-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001182123.1 | |
p53-regulated apoptosis-inducing protein 1 | |
Research Use Only | |
TP53AIP1 | |
Recombinant Protein | |
E. coli | |
MGSSSEVSFR SAQASCSGAR RQGLGRGDQN LSVMPPNGRA QTHTPGWVSP CSENRDGLLP ATAPGRLCSH RGADIPSFQT HQDPVTASGS SELHADCPQF RALDRAGN | |
Greater than 80% as determined by SDS-PAGE. |
63970 | |
200 μg | |
Not determined. | |
Full Length of BC069399 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |