missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OTUB1 / OTB1 Protein
A DNA sequence encoding the Homo sapiens (Human) OTUB1 / OTB1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8451-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_060140.2 | |
OTUB1 / OTB1 Protein | |
Research Use Only | |
OTUB1 | |
Recombinant Protein | |
E. coli | |
AAEEPQQQKQ EPLGSDSEGV NCLAYDEAIM AQQDRIQQEI AVQNPLVSER LELSVLYKEY AEDDNIYQQK IKDLHKKYSY IRKTRPDGNC FYRAFGFSHL EALLDDSKEL QRFKAVSAKS KEDLVSQGFT EFTIEDFHNT FMDLIEQVEK QTSVADLLAS FNDQSTSDYL VVYLRLLTSG YLQRESKFFE HFIEGGRTVK | |
Greater than 90% as determined by SDS-PAGE. |
55611 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |