missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OCM Protein
A cDNA sequence encoding the OCM was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12919-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
OCM Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS ITDVLSADDI S ITDVLSADDI AAALQECRDP DTFEPQKFFQ TSGLSKMSAN QVKDVFRFID NDQSGYLDEE ELKFFLQKFE SGARELTESE TKSLMAAADN DGDGKIGAEE FQEMVHS |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Oncomodulin-1 was filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20mM Tris and 50mM NaCl, pH 7.5. |