Learn More
enQuireBio™ Recombinant Human Obg-like ATPase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Obg-like ATPase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6451-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001011708.1 | |
Obg-like ATPase 1 Protein | |
Research Use Only | |
OLA1 | |
Recombinant Protein | |
E. coli | |
MPPKKGGDGI KPPPIIGRFG TSLKIGIVGL PNVGKSTFFN VLTNSQASAE NFPFCTIDPN ESRVPVPDER FDFLCQYHKP ASKIPAFLNV VDIAGLVKGA HNGQGLGNAF LSHISACDGI FHLTRAFEDD DITHVEGSVD PIRDIEIIHE ELQLKDEEMI GPIIDKLEKV AVRGGDKKLK PEYDIMCKVK SWVIDQKKPV RFYHDWNDKE IEVLNKHLFL TSKPMVYLVN LSEKDYIRKK NKWLIKIKEW VDKYDPGALV IPFSGALELK LQELSAEERQ KYLEANMTQS ALPKIIKAGF AALQLEYFFT AGPDEVRAWT IRKGTKAPQA AGKIHTDFEK GFIMAEVMKY EDFKEEGSEN AVKAAGKYRQ QGRNYIVEDG DIIFFKFNTP QQPKKK | |
Greater than 90% as determined by SDS-PAGE. |
29789 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |