missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolin Protein
A DNA sequence encoding the Homo sapiens (Human) Nucleolin, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP9301-ye-50ug
Weitere Details : Peso : 0.01000kg
Spezifikation
Nucleolin Protein | |
Research Use Only | |
NCL | |
Recombinant Protein | |
Yeast | |
VKLAKAGKNQ GDPKKMAPPP KEVEEDSEDE EMSEDEEDDS SGEEVVIPQK KGKKAAATSA KKVVVSPTKK VAVATPAKKA AVTPGKKAAA TPAKKTVTPA KAVTTPGKKG ATPGKALVAT PGKKGAAIPA KGAKNGKNAK KEDSDEEEDD DSEEDEEDDE DEDEDEDEIE PAAMKAAAAA PASEDEDDED DEDDEDGKSK GIAYIEFKTE ADAEKTFEEK QGTEIDGRSI SLYYTGEKGQ NQDYRGGKNS TWSGESKTLV LSNLSYSATE ETLQEVFEKA TFIKVPQNQN GKSKGYAFIE FASFEDAKEA LNSCNKREIE GRAIRLELQG PRGSPNARSQ PSKTLFVKGL SEDTTEETLK ESFDGSVRAR IVTDRETGSS KGFGFVDFNS EEDAKAAKEA MEDGEIDGNK VTLDWAKPKG EGGFGGRGGG RGGFGGRGGG RGGRGGFGGR GRGGFGGRGG FRGGRGGGGD HKPQGKKTKF E | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |