missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRN1 / Neuritin 1 Protein
A DNA sequence encoding the Human NRN1 / Neuritin 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10422-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001265639.1 | |
NRN1 / Neuritin 1 Protein | |
Research Use Only | |
NRN1 | |
The ED50 as determined by by a cell proliferation assay using rat C6 cells is less than 25 ng/ml, corresponding to a specific activity of > 4.0 M 104 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
51299 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
AGKCDAVFKG FSDCLLKLGD SMANYPQGLD DKTNIKTVCT YWEDFHSCTV TALTDCQEGA KDMWDKLRKE SKNLNIQGSL FELCGSGN | |
Greater than 97% as determined by SDS-PAGE and HPLC. |