Learn More
enQuireBio™ Recombinant Human NRAS Protein
A cDNA sequence encoding the NRAS was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12877-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NRAS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTEYKLVVVG AGGVGKSALT IQLIQNHFVD EYDPTIEDSY RKQVVIDGET CLLDILDTAG QEEYSAMRDQ YMRTGEGFLC VFAINNSKSF ADINLYREQI KRVKDSDDVP MVLVGNKCDL PTRTVDTKQA HELAKSYGIP FIETSAKTRQ GVEDAFYTLV REIRQYRMKK LNSSDDGTQG CMGLPC | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The NRAS protein solution contains 20mM Tris-HCl pH 7.5, 100mM NaCl, and 10% glycerol. |