missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NEK7 Protein
A cDNA sequence encoding the NEK7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12827-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NEK7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMDEQSQ GMQGPPVPQF QPQKALRPDM GYNTLANFRI EKKIGRGQFS EVYRAACLLD GVPVALKKVQ IFDLMDAKAR ADCIKEIDLL KQLNHPNVIK YYASFIEDNE LNIVLELADA GDLSRMIKHF KKQKRLIPER TVWKYFVQLC SALEHMHSRR VMHRDIKPAN VFITATGVVK LGDLGLGRFF SSKTTAAHSL VGTPYYMSPE RIHENGYNFK SDIWSLGCLL YEMAALQSPF YGDKMNLYSL CKKIEQCDYP PLPSDHYSEE LRQLVNMCIN PDPEKRPDVT YVYDVAKRMH ACTASS | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NEK7 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) , 0.15M NaCl and 20% glycerol. |