missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFB10 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFB10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6413-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004539.1 | |
NDUFB10 Protein | |
Research Use Only | |
NDUFB10 | |
Recombinant Protein | |
E. coli | |
MPDSWDKDVY PEPPRRTPVQ PNPIVYMMKA FDLIVDRPVT LVREFIERQH AKNRYYYYHR QYRRVPDITE CKEEDIMCMY EAEMQWKRDY KVDQEIINIM QDRLKACQQR EGQNYQQNCI KEVEQFTQVA KAYQDRYQDL GAYSSARKCL AKQRQRMLQE RKAAKEAAAA TS | |
Greater than 80% as determined by SDS-PAGE. |
4716 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |