missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAT6 Protein
A cDNA sequence encoding the NAT6 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12796-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
24142 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
NAT6 | |
Human | |
His | |
The NAT6 protein solution contains 20mM Tris-HCl, pH 8.0, 100mM NaCl and 20% Glycerol. |
NAT6 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQELTLSPGP AKLTPTLDPT HRMELILSTS PAELTLDPAC QPKLPLDSTC QPEMTFNPGP TELTLDPEHQ PEETPAPSLA ELTLEPVHRR PELLDACADL INDQWPRSRT SRLHSLGQSS DAFPLCLMLL SPHPTLEAAP VVVGHARLSR VLNQPQSLLV ETVVVARALR GRGFGRRLME GLEVFARARG FRKLHLTTHD QVHFYTHLGY QLGEPVQGLV FTSRRLPATL LNAFPTAPSP RPPRKAPNLT AQAAPRGPKG PPLPPPPPLP ECLTISPPVP SGPPSKSLLE TQYQNVRGRP IFWMEKDI |