missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAALADL2 Protein
A DNA sequence encoding the Homo sapiens (Human) NAALADL2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7674-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_996898.2 | |
NAALADL2 Protein | |
Research Use Only | |
NAALADL2 | |
Recombinant Protein | |
Yeast | |
GYYVHTNCPS DAPSSGTVDP QLYQEILKTI QAEDIKKSFR NLVQLYKNED DMEISKKIKT QWTSLGLEDV QFVNYSVLLD LPGPSPSTVT LSSSGQCFHP NGQPCSEEAR KDSSQDLLYS YAAYSAKGTL KAEVIDVSYG MADDLKRIRK IKNVTNQIAL LKLGKLPLLY KLSSLEKAGF GGVLLYIDPC DLPKTVNPSH DTFMVSLNPG GDPSTPGYPS VDESFRQSRS NLTSLLVQPI SAPLVAKLIS SPKARTKNEA CSSLELPNNE IRVVSMQVQT VTKLKTVTNV VGFVMGLTSP DRYIIVGSHH HTAHSYNGQE WASSTAIITA FIRALMSKVK RGWRPDRTIV FCSWGGTAFG NIGSYEWGED FKKVLQKNVV AYISLHSPIR GNSSLYPVAS PSLQQLVVEK NNFNCTRRAQ CPETNISSIQ IQGDADYFIN HLGVPIVQFA YEDIKTLEGP SFLSEARFST RATKIEEMDP SFNLHETITK LSGEVILQIA NEPVLPFNAL DIALEVQNNL KGDQPNTHQL LAMALRLRES AELFQSDEMR PANDPKERAP IRIRMLNDIL QDMEKSFLVK QAPPGFYRNI LYHLDEKTSR FSILIEAWEH CKPLASNETL QEALSEVLNS INSAQVYFKA GLDVFKSVLD GKN | |
Greater than 90% as determined by SDS-PAGE. |
254827 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |