missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N-acetylmuramoyl-L-alanine amidase Protein
A DNA sequence encoding the Homo sapiens (Human) N-acetylmuramoyl-L-alanine amidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7947-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_443122.3 | |
N-acetylmuramoyl-L-alanine amidase Protein | |
Research Use Only | |
PGLYRP2 | |
Recombinant Protein | |
E. coli | |
SLPLLMDSVI QALAELEQKV PAAKTRHTAS AWLMSAPNSG PHNRLYHFLL GAWSLNATEL DPCPLSPELL GLTKEVARHD VREGKEYGVV LAPDGSTVAV EPLLAGLEAG LQGRRVINLP LDSMAAPWET GDTFPDVVAI APDVRATSSP GLRDGSPDVT TADIGANTPD ATKGCPDVQA SLPDAKAKSP PTMVDSLLAV TLAGNLGLTF LRGSQTQSHP DLGTEGCWDQ LSAPRTFTLL DPKASLLTMA FLNGALDGVI LGDYLSRTPE PRPSLSHLLS QYYGAGVARD PGFRSNFRRQ NGAALTSASI LAQQVWGTLV LLQRLEPVHL QLQCMSQEQL AQVAANATKE FTEAFLGCPA IHPRCRWGAA PYRGRPKLLQ LPLGFLYVHH TYVPAPPCTD FTRCAANMRS MQRYHQDTQG WGDIGYSFVV GSDGYVYEGR GWHWVGAHTL GHNSRGFGVA IVGNYTAALP TEAALRTVRD TLPSCAVRAG LLRPDYALLG HRQLVRTDCP GDALFDLLRT WPHFTATVKP RPARSVSKRS RREPPPRTLP ATDLQ | |
Greater than 90% as determined by SDS-PAGE. |
114770 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |