missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MUC2 Protein
A DNA sequence encoding the Homo sapiens (Human) MUC2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6394-ye-50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
MUC2 Protein | |
Research Use Only | |
MUC2 | |
Recombinant Protein | |
Yeast | |
VCSTWGNFHY KTFDGDVFRF PGLCDYNFAS DCRGSYKEFA VHLKRGPGQA EAPAGVESIL LTIKDDTIYL TRHLAVLNGA VVSTPHYSPG LLIEKSDAYT KVYSRAGLTL MWNREDALML ELDTKFRNHT CGLCGDYNGL QSYSEFLSDG VLFSPLEFGN MQKINQPDVV CEDPEEEVAP ASCSEHRAEC ERLLTAEAFA DCQDL | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |