missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MOCS2 Protein
A cDNA sequence encoding the MOCS2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12715-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
4338 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSSL EISSSCFSLE TKLPLSPPLV EDSAFEPSRK DMDEVEEKSK DVINFTAEKL SVDEVSQLVI SPLCGAISLF VGTTRNNFEG KKVISLEYEA YLPMAENEVR KICSDIRQKW PVKHIAVFHR LGLVPVSEAS IIIAVSSAHR AASLEAVSYA IDTLKAKVPI WKKEIYEESS TWKGNKECFW ASNS | |
Greater than 90% as determined by SDS-PAGE. |
MOCS2 Protein | |
Research Use Only | |
MOCS2 | |
Human | |
His | |
The MOCS2 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 50% glycerol. |