missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MFAP2 Protein
A cDNA sequence encoding the MFAP2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12689-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MFAP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQGQYDL DPLPPFPDHV QYTHYSDQID NPDYYDYQEV TPRPSEEQFQ FQSQQQVQQE VIPAPTPEPG NAELEPTEPG PLDCREEQYP CTRLYSIHRP CKQCLNEVCF YSLRRVYVIN KEICVRTVCA HEELLRADLC RDKFSKCGVM ASSGLCQSVA ASCARSCGSC | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MFAP2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 20% glycerol. |