missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metallothionein-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Metallothionein-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8894-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005944.1 | |
Metallothionein-2 Protein | |
Research Use Only | |
MT2A | |
Recombinant Protein | |
E. coli | |
MDPNCSCAAG DSCTCAGSCK CKECKCTSCK KSCCSCCPVG CAKCAQGCIC KGASDKCSC | |
Greater than 90% as determined by SDS-PAGE. |
4502 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |