missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MCFD2 Protein
A cDNA sequence encoding the MCFD2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12660-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MCFD2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSHMEEPA ASFSQPGSMG LDKNTVHDQE HIMEHLEGVI NKPEAEMSPQ ELQLHYFKMH DYDGNNLLDG LELSTAITHV HKEEGSEQAP LMSEDELINI IDGVLRDDDK NNDGYIDYAE FAKSLQ | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The MCFD2 protein solution contains 20mM Tris-HCl, pH 7.5, 100mM NaCl and 10% glycerol. |