missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAD2L1 / MAD2 Protein
A DNA sequence encoding the Homo sapiens (Human) MAD2L1 / MAD2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7568-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002349.1 | |
MAD2L1 / MAD2 Protein | |
Research Use Only | |
MAD2L1 | |
Recombinant Protein | |
E. coli | |
ALQLSREQGI TLRGSAEIVA EFFSFGINSI LYQRGIYPSE TFTRVQKYGL TLLVTTDLEL IKYLNNVVEQ LKDWLYKCSV QKLVVVISNI ESGEVLERWQ FDIECDKTAK DDSAPREKSQ KAIQDEIRSV IRQITATVTF LPLLEVSCSF DLLIYTDKDL VVPEKWEESG PQFITNSEEV RLRSFTTTIH KVNSMVAYKI PVND | |
Greater than 90% as determined by SDS-PAGE. |
4085 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |