missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LY6G6D Protein
A DNA sequence encoding the Homo sapiens (Human) LY6G6D, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9258-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_067069.2 | |
LY6G6D Protein | |
Research Use Only | |
LY6G6D | |
Recombinant Protein | |
Yeast | |
NRMRCYNCGG SPSSSCKEAV TTCGEGRPQP GLEQIKLPGN PPVTLIHQHP ACVAAHHCNQ VETESVGDVT YPAHRDCYLG DLCNS | |
Greater than 90% as determined by SDS-PAGE. |
58530 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |