missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LSP1 Protein
A DNA sequence encoding the Homo sapiens (Human) LSP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6323-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001013271.1 | |
LSP1 Protein | |
Research Use Only | |
LSP1 | |
Recombinant Protein | |
E. coli | |
MAEASSDPGA EEREELLGPT AQWSVEDEEE AVHEQCQHER DRQLQAQDEE GGGHVPERPK QEMLLSLKPS EAPELDEDEG FGDWSQRPEQ RQQHEGAQGT LDSGEPPQCR SPEGEQEDRP GLHAYEKEDS DEVHLEELSL SKEGPGPEDT VQDNLGAAGA EEEQEEHQKC QQPRTPSPLV LEGTIEQSSP PLSPTTKLID RTESLNRSIE KSNSVKKSQP DLPISKIDQW LEQYTQAIET AGRTPKLARQ ASIELPSMAV ASTKSRWETG EVQAQSAAKT PSCKDIVAGD MSKKSLWEQK GGSKTSSTIK STPSGKRYKF VATGHGKYEK VLVEGGPAP | |
Greater than 80% as determined by SDS-PAGE. |
4046 | |
50 μg | |
Not determined. | |
Full Length of BC001785 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |