missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIM and SH3 domain protein 1
A DNA sequence encoding the Homo sapiens (Human) LIM and SH3 domain protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8479-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001258537.1 | |
LIM and SH3 domain protein 1 | |
Research Use Only | |
LASP1 | |
Recombinant Protein | |
E. coli | |
MNPNCARCGK IVYPTEKVNC LDKFWHKACF HCETCKMTLN MKNYKGYEKK PYCNAHYPKQ SFTMVADTPE NLRLKQQSEL QSQVRYKEEF EKNKGKGFSV VADTPELQRI KKTQDQISNI KYHEEFEKSR MGPSGGEGME PERRDSQDGS SYRRPLEQQQ PHHIPTSAPV YQQPQQQPVA QSYGGYKEPA APVSIQRSAP GGGGKRYRAV YDYSAADEDE VSFQDGDTIV NVQQIDDGWM YGT | |
Greater than 90% as determined by SDS-PAGE. |
3927 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |