missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leptin Protein
A DNA sequence encoding the Homo sapiens (Human) Leptin Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8559-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000221.1 | |
Leptin Protein | |
Research Use Only | |
LEP | |
Recombinant Protein | |
E. coli | |
DDTKTLIKTI VTRINDISHT QSVSSKQKVT GLDFIPGLHP ILTLSKMDQT LAVYQQILTS MPSRNVIQIS NDLENLRDLL HVLAFSKSCH LPWASGLETL DSLGGVLEAS GYSTEVVALS RLQGSLQDML WQLDLS | |
Greater than 90% as determined by SDS-PAGE. |
3952 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |