missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LEFTY1 Protein
A cDNA sequence encoding the LEFTY1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10746-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
LEFTY1 Protein | |
Research Use Only | |
The specific activity was determined by dose dependent ability to inhibit BMP-4 (6.5 ng/mL) induction of alkaline phosphatase production in the MC-3T3-E1 cell line (Mouse chondrogenic cell line) and is typically 10-40ng/ml. | |
Human | |
Untagged | |
The LEFTY1 was lyophilized from 1 mg/ml in 1xPBS. |
1 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
MQPLWLCWAL WVLPLASPGA ALTGEQLLGS LLRQLQLKEV PTLDRADMEE LVIPTHVRAQ YVALLQRSHG DRSRGKRFSQ SFREVAGRFL ALEASTHLLV FGMEQRLPPN SELVQAVLRL FQEPVPKAAL HRHGRLSPRS ARARVTVEWL RVRDDGSNRT SLIDSRLVSV HESGWKAFDV TEAVNFWQQL SRPRQPLLLQ VSVQREHLGP LASGAHKLVR FASQGAPAGL GEPQLELHTL DLGDYGAQGD CDPEAPMTEG TRCCRQEMYI DLQGMKWAEN WVLEPPGFLA YECVGTCRQP PEALAFKWPF LGPRQCIASE TDSLPMIVSI KEGGRTRPQV VSLPNMRVQK CSCASDGALV PRRLQP | |
Greater than 90% as obsereved by SDS-PAGE. |