missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KXD1 Protein
A cDNA sequence encoding the KXD1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12520-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
79036 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
KXD1 | |
Human | |
His | |
KXD1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
KXD1 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDLPDSA SRVFCGRILS MVNTDDVNAI ILAQKNMLDR FEKTNEMLLN FNNLSSARLQ QMSERFLHHT RTLVEMKRDL DSIFRRIRTL KGKLARQHPE AFSHIPEASF LEEEDEDPIP PSTTTTIATS EQSTGSCDTS PDTVSPSLSP GFEDLSHVQP GSPAINGRSQ TDDEEMTGE |