missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK1 / Kallikrein 1 Protein
A DNA sequence encoding the Homo sapiens (Human) KLK1 / Kallikrein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6266-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002248.1 | |
KLK1 / Kallikrein 1 Protein | |
Research Use Only | |
KLK1 | |
Recombinant Protein | |
E. coli | |
IVGGWECEQH SQPWQAALYH FSTFQCGGIL VHRQWVLTAA HCISDNYQLW LGRHNLFDDE NTAQFVHVSE SFPHPGFNMS LLENHTRQAD EDYSHDLMLL RLTEPADTIT DAVKVVELPT EEPEVGSTCL ASGWGSIEPE NFSFPDDLQC VDLKILPNDE CKKAHVQKVT DFMLCVGHLE GGKDTCVGDS GGPLMCDGVL QGVTSWGYVP CGTPNKPSVA VRVLSYVKWI EDTIAENS | |
Greater than 90% as determined by SDS-PAGE. |
3816 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |