missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IZUMO4 / C19orf36 Protein
A DNA sequence encoding the Homo sapiens (Human) IZUMO4 / C19orf36, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7632-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001026905.2 | |
IZUMO4 / C19orf36 Protein | |
Research Use Only | |
IZUMO4 | |
Recombinant Protein | |
E. coli | |
HGCLHCHSNF SKKFSFYRHH VNFKSWWVGD IPVSGALLTD WSDDTMKELH LAIPAKITRE KLDQVATAVY QMMDQLYQGK MYFPGYFPNE LRNIFREQVH LIQNAIIESR IDCQHRCGIF QYETISCNNC TDSHVACFGY NCESSAQWKS AVQGLLNYIN NWHKQDTSMS LVSPALRCLE PPHLANLTLE DAAECLKQH | |
Greater than 90% as determined by SDS-PAGE. |
113177 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |