missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ISOC2 Protein
A cDNA sequence encoding the ISOC2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12461-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ISOC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEQYPQGL GPTVPELGTE GLRPLAKTCF SMVPALQQEL DSRPQLRSVL LCGIEAQACI LNTTLDLLDR GLQVHVVVDA CSSRSQVDRL VALARMRQSG AFLSTSEGLI LQLVGDAVHP QFKEIQKLIK EPAPDSGLLG LFQGQNSLLH | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ISOC2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |