missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IRF 3 Protein
A cDNA sequence encoding the IRF 3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12458-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
3661 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGTPKPRILP WLVSQLDLGQ LEGVAWVNKS RTRFRIPWKH GLRQDAQQED FGIFQAWAEA TGAYVPGRDK PDLPTWKRNF RSALNRKEGL RLAEDRSKDP HDPHKIYEFV NS | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IRF 3 Protein | |
Research Use Only | |
IRF3 | |
Human | |
Untagged | |
1 mg/ml in phosphate-buffered saline (PBS), pH 7.4. |