missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IP-10 / CXCL10 Protein
A DNA sequence encoding the Homo sapiens (Human) IP-10 / CXCL10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8532-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001556.2 | |
IP-10 / CXCL10 Protein | |
Research Use Only | |
CXCL10 | |
Recombinant Protein | |
E. coli | |
VPLSRTVRCT CISISNQPVN PRSLEKLEII PASQFCPRVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP | |
Greater than 90% as determined by SDS-PAGE. |
3627 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |