missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha 10 / IFNA10 Protein
A DNA sequence encoding the Homo sapiens (Human) GDF-15, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5000-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004855.2 | |
Interferon alpha 10 / IFNA10 Protein | |
Research Use Only | |
GDF15 | |
Recombinant Protein | |
E. coli | |
RNGDHCPLGP GRCCRLHTVR ASLEDLGWAD WVLSPREVQV TMCIGACPSQ FRAANMHAQI KTSLHRLKPD TVPAPCCVPA SYNPMVLIQK TDTGVSLQTY DDLLAKDCHC I | |
Greater than 90% as determined by SDS-PAGE. |
9518 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |