missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL6 / Interleukin-6 Protein
A DNA sequence encoding the Human IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10313-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000591.1 | |
IL6 / Interleukin-6 Protein | |
Research Use Only | |
IL6 | |
Assay 1: The ED50 as determined by a cell proliferation assay using IL-6-dependent murine 7TD1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. Assay 2: The ED50 as determined by a cell proliferation assay using IL-6-dependent murine T1165 cells is less than 0.8 ng/ml, corresponding to a specific activity of > 1.25 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4. , lyophilized |
3569 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VPPGEDSKDV AAPHRQPLTS SERIDKQIRY ILDGISALRK ETCNKSNMCE SSKEALAENN LNLPKMAEK DGCFQSGFNE ETCLVKIITG LLEFEVYLEY LQNRFESSEE QARAVQMSTK VLIQFLQKK AKNLDAITTP DPTTNASLLT KLQAQNQWLQ DMTTHLILRS FKEFLQSSLR ALRQM | |
Greater than 96% as determined by SDS-PAGE and HPLC. |