missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL22 / Interleukin 22 Protein
Recombinant Protein
Marca: enQuireBio™ QP1166-1 mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_065386.1 | |
IL22 / Interleukin 22 Protein | |
Research Use Only | |
IL22 | |
Recombinant Protein | |
E. coli | |
MAPISSHCRL DKSNFQQPYI TNRTFMLAKE ASLADNNTDV RLIGEKLFHG VSMSERCYLM KQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQKLKDTV KKLGESGEIK AIGELDLLFM SLRNACI | |
Greater than 95% as determined by SDS-PAGE. |
50616 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human IL22 / Interleukin 22 induces pSTAT3 activation in HepG2 cells (detected via western blot at less than 5 ng/ml). | |
Human | |
Untagged | |
10 mM sodium citrate, pH 3.0 |