missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL22 / Interleukin 22 Protein
A DNA sequence encoding the Human IL22 / Interleukin 22, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10327-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_065386.1 | |
IL22 / Interleukin 22 Protein | |
Research Use Only | |
IL22 | |
The ED50 as determined by inducing IL-10 secretion of human COLO 205 cells is less than 0.3 ng/ml, corresponding to a specific activity of > 3.3 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 5.0, lyophilized |
50616 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MAPISSHCRL DKSNFQQPYI TNRTFMLAKE ASLADNNTDV RLIGEKLFHG VSMSERCYLM KQVLNFTLEE VLFPQSDRFQ PYMQEVVPFL ARLSNRLSTC HIEGDDLHIQ RNVQKLKDTV KKLGESGEIK AIGELDLLFM SLRNACI | |
Greater than 97% as determined by SDS-PAGE and HPLC. |