missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL20 / Interleukin-20 Protein
A DNA sequence encoding the Human IL20 / Interleukin-20, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10325-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_061194.2 | |
IL20 / Interleukin-20 Protein | |
Research Use Only | |
IL20 | |
The ED50 as determined by a cell proliferation assay using human IL-20Ralpha and human IL-20Rbeta co-transfected murine BaF3 pro-B cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.2, with trehalose, lyophilized |
50604 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE | |
Greater than 95% as determined by SDS-PAGE and HPLC. |