missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17F / IL-17F Protein
Recombinant Protein
Marca: enQuireBio™ QP1158-500 ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_443104.1 | |
IL17F / IL-17F Protein | |
Research Use Only | |
IL17F | |
Recombinant Protein | |
E. coli | |
MRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ | |
Greater than 95% as determined by SDS-PAGE. |
112744 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 100 ng/mL in the Production of IL-6 from mouse NIH 3T3 cells assay. The bioactivity acceptance criteria is set at 1 x 10^4 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |